Assay Details
| Gene Symbol: mea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002111.1 | NP_001002111.1 | 3 - 4 | 482 | 64 | |
| XM_005156496.4 | XP_005156553.1 | 3 - 4 | 519 | 64 | ||
| GenBank mRNA | BC071483.1 | - | 3 - 4 | 482 | 64 | |
More Information
Set Membership: |
|

mea1
Genomic Map