Assay Details
| Gene Symbol: tmprss4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077738.1 | NP_001071206.1 | 10 - 11 | 1103 | 126 | |
| XM_005157490.4 | XP_005157547.1 | 10 - 11 | 1386 | 126 | ||
| GenBank mRNA | BC125834.1 | - | 10 - 11 | 1103 | 126 | |
More Information
Set Membership: |
|

tmprss4a
Genomic Map