Assay Details
| Gene Symbol: spint1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111223.1 | NP_001104693.1 | 4 - 5 | 1093 | 65 | |
| GenBank mRNA | BC155551.1 | - | 4 - 5 | 1093 | 65 | |
| BC163929.1 | - | 4 - 5 | 976 | 65 | ||
| BC163937.1 | - | 4 - 5 | 976 | 65 | ||
| EF424430.1 | - | 4 - 5 | 1130 | 65 | ||
More Information
Set Membership: |
|

spint1b
Genomic Map