Assay Details
| Gene Symbol: spint1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111223.1 | NP_001104693.1 | 5 - 6 | 1124 | 77 | |
| GenBank mRNA | BC155551.1 | - | 5 - 6 | 1124 | 77 | |
| BC163929.1 | - | 5 - 6 | 1007 | 77 | ||
| BC163937.1 | - | 5 - 6 | 1007 | 77 | ||
| EF424430.1 | - | 5 - 6 | 1161 | 77 | ||
More Information
Set Membership: |
|

spint1b
Genomic Map