Assay Details
| Gene Symbol: phactr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114411.1 | NP_001107883.1 | 1 - 2 | 226 | 63 | |
| GenBank mRNA | BC154544.1 | - | 1 - 2 | 226 | 63 | |
More Information
Set Membership: |
|

phactr3a
Genomic Map