Assay Details
| Gene Symbol: SOCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123379.1 | NP_001116851.1 | 1 | 555 | 108 | |
| XM_023651604.1 | XP_023507372.1 | 860 | 108 | |||
| GenBank mRNA | EU554424.1 | - | 1 | 555 | 108 | |
More Information
Set Membership: |
|

SOCS3
Genomic Map