Assay Details
| Gene Symbol: LOC100071602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014731276.2 | XP_014586762.1 | 427 | 77 | ||
| XM_014731278.2 | XP_014586764.1 | 979 | 77 | |||
| XM_014731281.2 | XP_014586767.1 | 981 | 77 | |||
| XM_014731282.2 | XP_014586768.1 | 981 | 77 | |||
| XM_014731283.2 | XP_014586769.1 | 981 | 77 | |||
| XM_023619422.1 | XP_023475190.1 | 148 | 77 | |||
More Information
Set Membership: |
|

LOC100071602
Genomic Map