Assay Details
| Gene Symbol: RHOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005608809.3 | XP_005608866.1 | 420 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005608809.3 transcripts.
Set Membership: |
|

RHOH
Genomic Map