Assay Details
| Gene Symbol: RHOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003364726.4 | XP_003364774.1 | 324 | 96 | ||
| XM_005608809.3 | XP_005608866.1 | 542 | 96 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003364726.4, XM_005608809.3 transcripts.
Set Membership: |
|

RHOH
Genomic Map