Assay Details
| Gene Symbol: RPL7L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006452.1 | NP_001006452.1 | 2 - 3 | 151 | 108 | |
| GenBank mRNA | AJ720177.1 | - | 2 - 3 | 151 | 108 | |
More Information
Set Membership: |
|

RPL7L1
Genomic Map