Assay Details
| Gene Symbol: PLA2R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206983.1 | NP_996866.1 | 11 - 12 | 1849 | 60 | |
| GenBank mRNA | AY450642.1 | - | 11 - 12 | 1849 | 60 | |
More Information
Set Membership: |
|

PLA2R1
Genomic Map