Assay Details
| Gene Symbol: PTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031371.1 | NP_001026542.1 | 9 - 10 | 923 | 56 | |
| GenBank mRNA | AJ719528.1 | - | 9 - 10 | 923 | 56 | |
More Information
Set Membership: |
|

PTPA
Genomic Map