Assay Details
| Gene Symbol: NDUFB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006502.1 | NP_001006502.1 | 3 - 4 | 345 | 68 | |
| GenBank mRNA | AJ720322.1 | - | 3 - 4 | 345 | 68 | |
More Information
Set Membership: |
|

NDUFB8
Genomic Map