Assay Details
| Gene Symbol: SOCS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318450.1 | NP_001305379.1 | 1 | 424 | 106 | |
| XM_004937684.3 | XP_004937741.1 | 3 | 2599 | 106 | ||
| XM_004937685.3 | XP_004937742.1 | 3 | 1253 | 106 | ||
| XM_004937686.3 | XP_004937743.1 | 3 | 682 | 106 | ||
| XM_015289234.2 | XP_015144720.1 | 4 | 1537 | 106 | ||
| XM_015289239.2 | XP_015144725.1 | 4 | 1781 | 106 | ||
| XM_015289240.2 | XP_015144726.1 | 4 | 1374 | 106 | ||
| XM_015289242.2 | XP_015144728.1 | 4 | 835 | 106 | ||
| XM_015289248.2 | XP_015144734.1 | 3 | 689 | 106 | ||
| XM_015289257.2 | XP_015144743.1 | 3 | 1416 | 106 | ||
| XM_015289262.2 | XP_015144748.1 | 4 | 2717 | 106 | ||
| XM_015289270.2 | XP_015144756.1 | 4 | 2763 | 106 | ||
| XM_025148475.1 | XP_025004243.1 | 4 | 770 | 106 | ||
| GenBank mRNA | BX950848.2 | - | 1 | 471 | 106 | |
| HQ917699.1 | - | 1 | 424 | 106 | ||
More Information
Set Membership: |
|

SOCS1L
Genomic Map