Assay Details
| Gene Symbol: ATRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256151.1 | NP_001243080.1 | 12 - 13 | 2410 | 114 | |
| XM_015293045.2 | XP_015148531.1 | 11 - 12 | 2524 | 114 | ||
| XM_015293046.2 | XP_015148532.1 | 11 - 12 | 2521 | 114 | ||
| XM_015293047.2 | XP_015148533.1 | 11 - 12 | 2497 | 114 | ||
| GenBank mRNA | AB684452.1 | - | 12 - 13 | 2410 | 114 | |
More Information
Set Membership: |
|

ATRIP
Genomic Map