Assay Details
| Gene Symbol: LOC101749151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293893.2 | XP_015149379.1 | 13 - 14 | 1189 | 91 | |
| XM_015293894.1 | XP_015149380.1 | 13 - 14 | 1313 | 91 | ||
| XM_025154972.1 | XP_025010740.1 | 13 - 14 | 1173 | 91 | ||
More Information
Set Membership: |
|

LOC101749151
Genomic Map