Assay Details
| Gene Symbol: LOC101749151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293894.1 | XP_015149380.1 | 1 - 2 | 177 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015293894.1 transcripts.
Set Membership: |
|

LOC101749151
Genomic Map