Assay Details
| Gene Symbol: LOC101749151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293893.2 | XP_015149379.1 | 7 - 8 | 710 | 69 | |
| XM_015293894.1 | XP_015149380.1 | 7 - 8 | 834 | 69 | ||
| XM_025154972.1 | XP_025010740.1 | 7 - 8 | 694 | 69 | ||
More Information
Set Membership: |
|

LOC101749151
Genomic Map