Assay Details
| Gene Symbol: RSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297035.2 | XP_015152521.1 | 5 - 6 | 604 | 108 | |
| XM_025142658.1 | XP_024998426.1 | 4 - 5 | 779 | 108 | ||
| GenBank mRNA | BX931751.1 | - | 4 - 5 | 525 | 108 | |
More Information
Set Membership: |
|

RSG1
Genomic Map