Assay Details
| Gene Symbol: Asic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001279840.1 | NP_001266769.1 | 3 - 4 | 950 | 91 | |
| XM_013078965.2 | XP_012934419.1 | 5 - 6 | 931 | 91 | ||
| GenBank mRNA | JF912493.1 | - | 4 - 5 | 839 | 91 | |
| JN703734.1 | - | 3 - 4 | 950 | 91 | ||
More Information
Set Membership: |
|

Asic1
Genomic Map