Assay Details
| Gene Symbol: Plpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838814.2 | XP_004838871.1 | 4 - 5 | 455 | 79 | |
| XM_021253332.1 | XP_021108991.1 | 6 - 7 | 1346 | 79 | ||
| XM_021253333.1 | XP_021108992.1 | 4 - 5 | 503 | 79 | ||
| XM_021253334.1 | XP_021108993.1 | 5 - 6 | 1079 | 79 | ||
More Information
Set Membership: |
|

Plpp4
Genomic Map