Assay Details
| Gene Symbol: Asic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001279840.1 | NP_001266769.1 | 7 - 8 | 1398 | 71 | |
| XM_013078965.2 | XP_012934419.1 | 9 - 10 | 1379 | 71 | ||
| XM_021243007.1 | XP_021098666.1 | 9 - 10 | 1382 | 71 | ||
| GenBank mRNA | JF912493.1 | - | 8 - 9 | 1287 | 71 | |
| JN703734.1 | - | 7 - 8 | 1398 | 71 | ||
More Information
Set Membership: |
|

Asic1
Genomic Map