Assay Details
| Gene Symbol: LOC101702191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262275.1 | XP_021117934.1 | 4 - 5 | 586 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021262275.1 transcripts.
Set Membership: |
|

LOC101702191
Genomic Map