Assay Details
| Gene Symbol: LOC101713485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872186.3 | XP_004872243.1 | 7 - 8 | 1187 | 61 | |
| XM_004872187.3 | XP_004872244.1 | 7 - 8 | 1123 | 61 | ||
| XM_004872188.3 | XP_004872245.1 | 7 - 8 | 1188 | 61 | ||
More Information
Set Membership: |
|

LOC101713485
Genomic Map