Assay Details
| Gene Symbol: Rpl7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846561.3 | XP_004846618.1 | 5 - 6 | 794 | 60 | |
| XM_004846562.3 | XP_004846619.1 | 5 - 6 | 857 | 60 | ||
More Information
Set Membership: |
|

Rpl7l1
Genomic Map