Assay Details
| Gene Symbol: LOC101711537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262276.1 | XP_021117935.1 | 2 - 3 | 120 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021262276.1 transcripts.
Set Membership: |
|

LOC101711537
Genomic Map