Assay Details
| Gene Symbol: LOC101711537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262276.1 | XP_021117935.1 | 8 - 9 | 871 | 81 | |
| XM_021262277.1 | XP_021117936.1 | 6 - 7 | 680 | 81 | ||
| XM_021262278.1 | XP_021117937.1 | 6 - 7 | 868 | 81 | ||
More Information
Set Membership: |
|

LOC101711537
Genomic Map