Assay Details
| Gene Symbol: Spint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861586.2 | XP_004861643.1 | 10 - 11 | 1767 | 67 | |
| XM_021241849.1 | XP_021097508.1 | 10 - 11 | 1451 | 67 | ||
More Information
Set Membership: |
|

Spint1
Genomic Map