Assay Details
| Gene Symbol: Milr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870229.3 | XP_004870286.2 | 6 - 7 | 840 | 83 | |
| XM_021248648.1 | XP_021104307.1 | 7 - 8 | 848 | 83 | ||
| XM_021248649.1 | XP_021104308.1 | 6 - 7 | 726 | 83 | ||
| XM_021248650.1 | XP_021104309.1 | 7 - 8 | 918 | 83 | ||
| XM_021248651.1 | XP_021104310.1 | 6 - 7 | 744 | 83 | ||
| XM_021248652.1 | XP_021104311.1 | 6 - 7 | 744 | 83 | ||
| XM_021248653.1 | XP_021104312.1 | 7 - 8 | 733 | 83 | ||
| XM_021248654.1 | XP_021104313.1 | 6 - 7 | 582 | 83 | ||
More Information
Set Membership: |
|

Milr1
Genomic Map