Assay Details
| Gene Symbol: PRSS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031948.4 | NP_114154.1 | 1 - 2 | 442 | 100 | |
| GenBank mRNA | AB056161.1 | - | 1 - 2 | 101 | 100 | |
| AJ306593.1 | - | 1 - 2 | 70 | 100 | ||
| AY359106.1 | - | 1 - 2 | 86 | 100 | ||
| BC034294.2 | - | 1 - 2 | 106 | 4752547 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000789.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB056161.1,AJ306593.1,AY359106.1,BC034294.2,NM_031948.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011576.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011576.hg.1 | TC16000789.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011575.hg.1 | ENST00000302641.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011575.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011575.hg.1 | TC16000789.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011574.hg.1 | ENST00000302641.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011574.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011574.hg.1 | TC16000789.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | TC1600009134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169549.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000543965.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | ENST00000566492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009134.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169551.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169551.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169551.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169551.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169551.hg.1 | TC1600009134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600169551.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169551.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169551.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169551.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169552.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169552.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169552.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169552.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169552.hg.1 | TC1600009134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600169552.hg.1 | ENST00000302641.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169552.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169552.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169552.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169553.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169553.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169553.hg.1 | NM_031948.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169553.hg.1 | TC1600009134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600169553.hg.1 | ENST00000562249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169553.hg.1 | ENST00000565903.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169553.hg.1 | NM_031948.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 162550 | - | - |

PRSS27
Genomic Map