Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105370619 | LOC105370619 | -42.6kb | Sense | Chr. 14 - 90515806 - 90517209 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928909 | LOC101928909 | 80.6kb | Anti-sense | Chr. 14 - 90642629 - 90648891 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map