| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100009707.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100173301.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173301.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100173301.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173301.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100173301.hg.1 |
TC1100009707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100173302.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173302.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100173302.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173302.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100173302.hg.1 |
TC1100009707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100173303.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173303.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173303.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100173303.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173303.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173303.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100173303.hg.1 |
TC1100009707.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100173304.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173304.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173304.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100173304.hg.1 |
ENST00000322028.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173304.hg.1 |
ENST00000534030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100173304.hg.1 |
NM_021128.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100173304.hg.1 |
TC1100009707.hg.1 |
Clariom D TC |