Assay Details
| Gene Symbol: MRPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145255.3 | NP_660298.2 | 3 - 4 | 413 | 85 | |
| NM_148887.2 | NP_683685.1 | 4 - 5 | 511 | 85 | ||
| NR_037575.1 | 3 - 4 | 562 | 85 | |||
| GenBank mRNA | AK127167.1 | - | 3 - 4 | 1288 | 85 | |
| AK293624.1 | - | 3 - 4 | 551 | 85 | ||
| AL558270.3 | - | 4 - 5 | 476 | 85 | ||
| AL701268.1 | - | 3 - 4 | 410 | 85 | ||
| BC015904.1 | - | 3 - 4 | 3919892 | 85 | ||
| BC039852.1 | - | 3 - 4 | 392 | 85 | ||
| BC052601.1 | - | 3 - 4 | 402 | 6085551 | 85 | |
| DA137043.1 | - | 3 - 4 | 413 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001627.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17021797.hg.1 | ENST00000290208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021797.hg.1 | ENST00000351111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021797.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021797.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021797.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021797.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021797.hg.1 | TC17001627.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000480901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000480901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000480901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | ENST00000480901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021799.hg.1 | ENST00000290208.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021799.hg.1 | ENST00000351111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021799.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021799.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021799.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021799.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17021799.hg.1 | TC17001627.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | TC1700010960.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188123.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | TC1700010960.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700188125.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000290208.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000351111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000414011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000421763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000423147.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000466016.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000480901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000480901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000480901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | ENST00000480901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_145255.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NM_148887.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010960.hg.1 | NR_037575.1 | Refseq |

MRPL10
Genomic Map