Assay Details
| Gene Symbol: PALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014431.2 | NP_055246.2 | 1 - 2 | 253 | 116 | |
| GenBank mRNA | AB033100.2 | - | 1 - 2 | 251 | 116 | |
| BC136375.1 | - | 1 - 2 | 135 | 9020362 | 116 | |
| BC136376.1 | - | 1 - 2 | 135 | 9020363 | 116 | |
| CN335744.1 | - | 1 - 2 | 278 | 116 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000430.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB033100.2,BC136375.1,BC136376.1,NM_014431.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10005023.hg.1 | ENST00000263563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005023.hg.1 | ENST00000373214.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005023.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005023.hg.1 | TC10000430.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005024.hg.1 | ENST00000263563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005024.hg.1 | ENST00000373214.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005024.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005024.hg.1 | TC10000430.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005022.hg.1 | ENST00000263563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005022.hg.1 | ENST00000373214.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005022.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10005022.hg.1 | TC10000430.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153787.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153787.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153787.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153787.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153787.hg.1 | TC1000007930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000153788.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153788.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153788.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153788.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153788.hg.1 | TC1000007930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000153789.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153789.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153789.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153789.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153789.hg.1 | TC1000007930.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007930.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153790.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153790.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153790.hg.1 | ENST00000263563.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000153790.hg.1 | NM_014431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000153790.hg.1 | TC1000007930.hg.1 | Clariom D TC |

PALD1
Genomic Map