Assay Details
| Gene Symbol: ASIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182847.2 | NP_878267.2 | 5 - 6 | 1729 | 65 | |
| XM_005246671.2 | XP_005246728.2 | 6 - 7 | 1643 | 65 | ||
| XM_017004439.1 | XP_016859928.1 | 6 - 7 | 802 | 65 | ||
| GenBank mRNA | BC010439.1 | - | 5 - 6 | 1569 | 4215164 | 65 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001330.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02020831.hg.1 | TC02001330.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020831.hg.1 | ENST00000347842.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020831.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020831.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020832.hg.1 | TC02001330.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020832.hg.1 | ENST00000347842.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020832.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02020832.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177796.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177796.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177796.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177796.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177796.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177796.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177796.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177796.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177796.hg.1 | TC0200010853.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000461395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000473709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010853.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177795.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177795.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177795.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177795.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177795.hg.1 | ENST00000347842.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177795.hg.1 | ENST00000358078.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177795.hg.1 | ENST00000474489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200177795.hg.1 | NM_182847.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200177795.hg.1 | TC0200010853.hg.1 | Clariom D TC |

ASIC4
Genomic Map