Assay Details
| Gene Symbol: GRASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181711.3 | NP_859062.1 | 2 - 3 | 369 | 90 | |
| GenBank mRNA | BC053666.1 | - | 2 - 3 | 350 | 6151168 | 90 |
| HY052304.1 | - | 2 - 3 | 370 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154482.hg.1 | TC1200007652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |

GRASP
Genomic Map