Assay Details
| Gene Symbol: MTUS2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046378.1 | 1 - 2 | 256 | 90 | ||
| GenBank mRNA | BC027486.1 | - | 1 - 2 | 256 | 90 | |
| BI223246.1 | - | 1 - 2 | 255 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000626029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | ENST00000626029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000626029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | ENST00000626029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008485.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157188.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157188.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157188.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157188.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157188.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157188.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157188.hg.1 | TC1300008485.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000626029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000323380.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000587588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | ENST00000626029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | NR_046378.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157185.hg.1 | TC1300008485.hg.1 | Clariom D TC |

MTUS2-AS1
Genomic Map