Assay Details
| Gene Symbol: CEACAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002483.6 | NP_002474.4 | 4 - 5 | 1074 | 62 | |
| GenBank mRNA | AK312542.1 | - | 4 - 5 | 1059 | 62 | |
| BC005008.1 | - | 4 - 5 | 995 | 3640231 | 62 | |
| BT009774.1 | - | 4 - 5 | 955 | 62 | ||
| M18216.1 | - | 4 - 5 | 1001 | 62 | ||
| M18728.1 | - | 4 - 5 | 1005 | 62 | ||
| M29541.1 | - | 4 - 5 | 1048 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000584.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19008117.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008117.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008117.hg.1 | TC19000584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008115.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008115.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008115.hg.1 | TC19000584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008120.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008120.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008120.hg.1 | TC19000584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010840.hg.1 | ENST00000457676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010840.hg.1 | ENST00000457676.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008118.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008118.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008118.hg.1 | TC19000584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008119.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008119.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008119.hg.1 | TC19000584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008116.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008116.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008116.hg.1 | TC19000584.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900201136.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201136.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201136.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201136.hg.1 | TC1900011742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201136.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201136.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201136.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201137.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201137.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201137.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201137.hg.1 | TC1900011742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201137.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201137.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201137.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011742.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010811.hg.1 | ENST00000434624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201135.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201135.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201135.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201135.hg.1 | TC1900011742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201135.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201135.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201135.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201142.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201142.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201142.hg.1 | TC1900011742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201142.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201142.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010840.hg.1 | ENST00000457676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010840.hg.1 | ENST00000457676.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201139.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201139.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201139.hg.1 | TC1900011742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201139.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201139.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201141.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201141.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201141.hg.1 | TC1900011742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201141.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201141.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201138.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201138.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201138.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201138.hg.1 | TC1900011742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201138.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201138.hg.1 | ENST00000595740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201138.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201140.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201140.hg.1 | NM_002483.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900201140.hg.1 | TC1900011742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900201140.hg.1 | ENST00000199764.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900201140.hg.1 | NM_002483.6 | Refseq |

CEACAM6
Genomic Map