Assay Details
| Gene Symbol: GRASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181711.3 | NP_859062.1 | 4 - 5 | 526 | 91 | |
| XM_005268691.3 | XP_005268748.1 | 4 - 5 | 1694 | 91 | ||
| XM_011537996.1 | XP_011536298.1 | 3 - 4 | 1472 | 91 | ||
| GenBank mRNA | BC052967.1 | - | 3 - 4 | 210 | 91 | |
| BC053666.1 | - | 4 - 5 | 507 | 6151168 | 91 | |
| HY035281.1 | - | 3 - 4 | 226 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000413.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005281.hg.1 | ENST00000293662.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005281.hg.1 | ENST00000380039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005281.hg.1 | ENST00000552049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005281.hg.1 | ENST00000552963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005281.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005281.hg.1 | TC12000413.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000550809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007652.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000293662.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000546756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000547749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000548505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000552049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000552508.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | ENST00000552963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | NM_001271856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | NM_181711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200154491.hg.1 | TC1200007652.hg.1 | Clariom D TC |

GRASP
Genomic Map