Assay Details
| Gene Symbol: PRSS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003619.3 | NP_003610.2 | 2 - 3 | 683 | 96 | |
| XM_005263318.4 | XP_005263375.1 | 2 - 3 | 956 | 96 | ||
| XM_011532387.2 | XP_011530689.1 | 2 - 3 | 958 | 96 | ||
| GenBank mRNA | AJ001531.1 | - | 2 - 3 | 676 | 96 | |
| BC007761.2 | - | 2 - 3 | 683 | 4126901 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001494.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04021175.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021175.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04021175.hg.1 | TC04001494.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000510903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000510903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000510903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180248.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180248.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180248.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400180248.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400180248.hg.1 | TC0400011685.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000296498.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000503043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000510903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000510903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000510903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | ENST00000515089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011685.hg.1 | NM_003619.3 | Refseq |

PRSS12
Genomic Map