Assay Details
| Gene Symbol: PHACTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199505.1 | NP_001186434.1 | 9 - 10 | 1368 | 66 | |
| NM_001199506.1 | NP_001186435.1 | 9 - 10 | 1363 | 66 | ||
| NM_001281507.1 | NP_001268436.1 | 9 - 10 | 1467 | 66 | ||
| NM_080672.4 | NP_542403.1 | 9 - 10 | 1845 | 66 | ||
| NM_183244.1 | NP_899067.1 | 9 - 10 | 1488 | 66 | ||
| NM_183246.1 | NP_899069.1 | 8 - 9 | 1278 | 66 | ||
| XM_011528525.2 | XP_011526827.1 | 9 - 10 | 1474 | 66 | ||
| XM_011528526.2 | XP_011526828.1 | 8 - 9 | 1280 | 66 | ||
| XM_017027626.1 | XP_016883115.1 | 9 - 10 | 1577 | 66 | ||
| XM_017027627.1 | XP_016883116.1 | 9 - 10 | 1522 | 66 | ||
| XM_017027628.1 | XP_016883117.1 | 8 - 9 | 1170 | 66 | ||
| XM_017027629.1 | XP_016883118.1 | 8 - 9 | 1313 | 66 | ||
| XM_017027630.1 | XP_016883119.1 | 8 - 9 | 1141 | 66 | ||
| XM_017027631.1 | XP_016883120.1 | 9 - 10 | 1404 | 66 | ||
| GenBank mRNA | AB098521.1 | - | 9 - 10 | 1488 | 66 | |
| AB098522.1 | - | 8 - 9 | 1278 | 66 | ||
| AJ311122.2 | - | 9 - 10 | 1844 | 66 | ||
| AJ617581.1 | - | 9 - 10 | 1363 | 66 | ||
| AK091632.1 | - | 9 - 10 | 1490 | 66 | ||
| AK098788.1 | - | 9 - 10 | 1368 | 66 | ||
| AK295459.1 | - | 9 - 10 | 1467 | 66 | ||
| AK314493.1 | - | 9 - 10 | 1575 | 66 | ||
| AK316047.1 | - | 9 - 10 | 1360 | 66 | ||
| BC108303.1 | - | 9 - 10 | 1845 | 4179728 | 66 | |
| BC117362.1 | - | 9 - 10 | 1733 | 40125913 | 66 | |
| BC117364.1 | - | 9 - 10 | 1736 | 40125915 | 66 | |
More Information
Set Membership: |
|

PHACTR3
Genomic Map