| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009364.hg.1 |
ENST00000258180.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009364.hg.1 |
ENST00000538274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009364.hg.1 |
NM_014732.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16009364.hg.1 |
TC16000661.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000258180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000538274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000562564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000562580.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000566428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
NM_001286565.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
NM_001286566.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
NM_014732.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
TC1600008675.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000258180.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000538274.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000562564.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000562580.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
ENST00000566428.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
NM_001286565.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
NM_001286566.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600164872.hg.1 |
NM_014732.3 |
Refseq |