Assay Details
| Gene Symbol: CEACAM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039213.3 | NP_001034302.2 | 3 - 4 | 588 | 95 | |
| XM_017026795.1 | XP_016882284.1 | 2 - 3 | 1283 | 95 | ||
| GenBank mRNA | BC144608.1 | - | 3 - 4 | 588 | 95 | |
| EU021223.1 | - | 2 - 3 | 382 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000626.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008679.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008679.hg.1 | TC19000626.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008686.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008686.hg.1 | TC19000626.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008689.hg.1 | ENST00000396750.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008689.hg.1 | ENST00000405314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008689.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008689.hg.1 | TC19000626.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008691.hg.1 | ENST00000396750.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008691.hg.1 | ENST00000405314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008691.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008691.hg.1 | TC19000626.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008690.hg.1 | ENST00000396750.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008690.hg.1 | ENST00000405314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008690.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008690.hg.1 | TC19000626.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008680.hg.1 | ENST00000396750.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008680.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19008680.hg.1 | TC19000626.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900163198.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163198.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163198.hg.1 | TC1900008276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163198.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163198.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008276.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163204.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163204.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163204.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163204.hg.1 | TC1900008276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163204.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163204.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163204.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163197.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163197.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163197.hg.1 | TC1900008276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163197.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163197.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163203.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163203.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163203.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163203.hg.1 | TC1900008276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163203.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163203.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163203.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163206.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163206.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163206.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163206.hg.1 | TC1900008276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163206.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163206.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163206.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163205.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163205.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163205.hg.1 | NM_001039213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900163205.hg.1 | TC1900008276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900163205.hg.1 | ENST00000405314.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163205.hg.1 | ENST00000587331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900163205.hg.1 | NM_001039213.3 | Refseq |

CEACAM16
Genomic Map