Assay Details
| Gene Symbol: CHUK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278.4 | NP_001269.3 | 20 - 21 | 2292 | 81 | |
| NM_001320928.1 | NP_001307857.1 | 20 - 21 | 2275 | 81 | ||
| GenBank mRNA | AB209028.1 | - | 19 - 20 | 4575 | 81 | |
| AF080157.1 | - | 20 - 21 | 2309 | 81 | ||
| AL133012.1 | - | 2 - 3 | 3591 | 81 | ||
| BC092514.1 | - | 20 - 21 | 2230 | 3063613 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10002931.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
stimulatory C-type lectin receptor signaling pathway
skeletal muscle contraction
protein phosphorylation
inflammatory response
immune response
I-kappaB kinase/NF-kappaB signaling
I-kappaB phosphorylation
Rho protein signal transduction
response to virus
response to toxic substance
anatomical structure morphogenesis
response to acetate
regulation of tumor necrosis factor-mediated signaling pathway
response to lipopolysaccharide
response to hydroperoxide
TRIF-dependent toll-like receptor signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
response to drug
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to amino acid
innate immune response
positive regulation of transcription from RNA polymerase II promoter
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
striated muscle cell differentiation
stress-activated MAPK cascade
nucleotide-binding oligomerization domain containing signaling pathway
cellular response to tumor necrosis factor
protein kinase activity
protein binding
ATP binding
IkappaB kinase activity
protein homodimerization activity
protein heterodimerization activity
scaffold protein binding
stimulatory C-type lectin receptor signaling pathway
skeletal muscle contraction
protein phosphorylation
inflammatory response
immune response
I-kappaB kinase/NF-kappaB signaling
I-kappaB phosphorylation
Rho protein signal transduction
response to virus
response to toxic substance
anatomical structure morphogenesis
response to acetate
regulation of tumor necrosis factor-mediated signaling pathway
response to lipopolysaccharide
response to hydroperoxide
TRIF-dependent toll-like receptor signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
response to drug
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to amino acid
innate immune response
positive regulation of transcription from RNA polymerase II promoter
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
striated muscle cell differentiation
stress-activated MAPK cascade
nucleotide-binding oligomerization domain containing signaling pathway
cellular response to tumor necrosis factor
protein kinase activity
protein binding
ATP binding
IkappaB kinase activity
protein homodimerization activity
protein heterodimerization activity
scaffold protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000585551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000585551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000585551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000585551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020469.hg.1 | ENST00000370397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020469.hg.1 | ENST00000485693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020469.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020469.hg.1 | TC10002931.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020468.hg.1 | ENST00000370397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020468.hg.1 | ENST00000485693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020468.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020468.hg.1 | TC10002931.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000187767.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187767.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187767.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187767.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187767.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187767.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187767.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187767.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187767.hg.1 | TC1000012585.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000585551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000585551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000585551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000585551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012585.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187766.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187766.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187766.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187766.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187766.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187766.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187766.hg.1 | TC1000012585.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000187768.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187768.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187768.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187768.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187768.hg.1 | ENST00000370397.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187768.hg.1 | ENST00000588656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187768.hg.1 | ENST00000590930.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000187768.hg.1 | NM_001278.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000187768.hg.1 | TC1000012585.hg.1 | Clariom D TC |

CHUK
Genomic Map