Assay Details
| Gene Symbol: TRAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032271.2 | NP_115647.2 | 15 - 16 | 1486 | 70 | |
| XM_005255627.4 | XP_005255684.1 | 15 - 16 | 1526 | 70 | ||
| XM_011522700.1 | XP_011521002.1 | 15 - 16 | 1528 | 70 | ||
| GenBank mRNA | AK090624.1 | - | 15 - 16 | 1500 | 70 | |
| AL136921.1 | - | 14 - 15 | 1382 | 70 | ||
| AY569455.1 | - | 14 - 15 | 1439 | 70 | ||
| BC024267.1 | - | 13 - 14 | 1272 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000063.hg.1
Set Membership: |
|

TRAF7
Genomic Map