Assay Details
| Gene Symbol: TRAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032271.2 | NP_115647.2 | 7 - 8 | 578 | 77 | |
| XM_005255627.4 | XP_005255684.1 | 7 - 8 | 624 | 77 | ||
| XM_011522700.1 | XP_011521002.1 | 7 - 8 | 620 | 77 | ||
| GenBank mRNA | AK090624.1 | - | 7 - 8 | 592 | 77 | |
| AL136921.1 | - | 6 - 7 | 474 | 77 | ||
| AY569455.1 | - | 6 - 7 | 531 | 77 | ||
| BC024267.1 | - | 5 - 6 | 364 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000063.hg.1
Set Membership: |
|

TRAF7
Genomic Map