Assay Details
| Gene Symbol: TRAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032271.2 | NP_115647.2 | 8 - 9 | 764 | 66 | |
| XM_005255627.4 | XP_005255684.1 | 8 - 9 | 810 | 66 | ||
| XM_011522700.1 | XP_011521002.1 | 8 - 9 | 806 | 66 | ||
| GenBank mRNA | AK090624.1 | - | 8 - 9 | 778 | 66 | |
| AL136921.1 | - | 7 - 8 | 660 | 66 | ||
| AY569455.1 | - | 7 - 8 | 717 | 66 | ||
| BC024267.1 | - | 6 - 7 | 550 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000063.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKKK activity
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
protein ubiquitination
positive regulation of MAPK cascade
positive regulation of apoptotic signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
activation of MAPKKK activity
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
protein ubiquitination
positive regulation of MAPK cascade
positive regulation of apoptotic signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001303.hg.1 | ENST00000326181.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001303.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001303.hg.1 | TC16000063.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600146337.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146337.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146337.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600146337.hg.1 | TC1600006594.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600146337.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146337.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146337.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000326181.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000564067.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000565383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567645.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000567653.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000569686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | ENST00000570169.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006594.hg.1 | NM_032271.2 | Refseq |

TRAF7
Genomic Map