Assay Details
| Gene Symbol: CADM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127173.1 | NP_001120645.1 | 4 - 5 | 661 | 82 | |
| NM_021189.3 | NP_067012.1 | 5 - 6 | 800 | 82 | ||
| GenBank mRNA | AF062733.2 | - | 5 - 6 | 757 | 82 | |
| AF363367.1 | - | 4 - 5 | 659 | 82 | ||
| AK001560.1 | - | 1 - 2 | 59 | 82 | ||
| AK289541.1 | - | 5 - 6 | 800 | 82 | ||
| AY046418.1 | - | 4 - 5 | 564 | 82 | ||
| AY358332.1 | - | 4 - 5 | 665 | 82 | ||
| BC033819.1 | - | 4 - 5 | 609 | 5199627 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006336.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK001560.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
protein localization
adherens junction organization
receptor activity
receptor binding
protein homodimerization activity
cell adhesion molecule binding
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
protein localization
adherens junction organization
receptor activity
receptor binding
protein homodimerization activity
cell adhesion molecule binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01021167.hg.1 | ENST00000368124.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021167.hg.1 | ENST00000368125.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021167.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021167.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021167.hg.1 | TC01006336.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021168.hg.1 | ENST00000368124.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021168.hg.1 | ENST00000368125.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021168.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021168.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01021168.hg.1 | TC01006336.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239752.hg.1 | TC0100018306.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | ENST00000416746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018306.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239753.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239753.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239753.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239753.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239753.hg.1 | ENST00000368124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239753.hg.1 | ENST00000368125.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100239753.hg.1 | NM_001127173.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239753.hg.1 | NM_021189.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100239753.hg.1 | TC0100018306.hg.1 | Clariom D TC |

CADM3
Genomic Map