| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10005030.hg.1 |
ENST00000263563.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10005030.hg.1 |
ENST00000373214.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10005030.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10005030.hg.1 |
TC10000430.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000153796.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153796.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000153796.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000153796.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000153796.hg.1 |
TC1000007930.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
ENST00000263563.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007930.hg.1 |
NM_014431.2 |
Refseq |