Assay Details
| Gene Symbol: PRSS56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195129.1 | NP_001182058.1 | 6 - 7 | 848 | 69 | |
| GenBank mRNA | BI464671.1 | - | 3 - 4 | 402 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001401.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3345309 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P15182 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022115.hg.1 | TC02001401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022115.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022114.hg.1 | TC02001401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02022114.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179785.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179785.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179785.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179785.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179785.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179785.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179785.hg.1 | TC0200011103.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011103.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179783.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179783.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179783.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179783.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179783.hg.1 | ENST00000449534.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179783.hg.1 | ENST00000602410.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179783.hg.1 | ENST00000617714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179783.hg.1 | NM_001195129.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179783.hg.1 | TC0200011103.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017103 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017103 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 105205 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 146592 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1674447 | NM_001195129.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1674447 | CN280933.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1674447 | NM_001195129.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1674447 | CN280933.1 | human_transcriptome |

PRSS56
Genomic Map